Learn More
Medchemexpress LLC HY-B1596 50mg Medchemexpress, Ceftizoxime CAS:68401-81-0 Purity:>98%
SDP
Catalog No. 501932974
Encompass_Preferred
Medchemexpress, HY-B1596 50mg Ceftizoxime CAS:68401-81-0 Ceftizoxime is a bacterial inhibitor which acts by interfering with bacterial cell wall synthesis and inhibiting cross-linking of the peptidoglycan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2974 Supplier Medchemexpress LLC Supplier No. HYB159650MG
Medchemexpress, HY-B1596 50mg Ceftizoxime CAS:68401-81-0 Ceftizoxime is a bacterial inhibitor which acts by interfering with bacterial cell wall synthesis and inhibiting cross-linking of the peptidoglycan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.