Learn More
Medchemexpress LLC HY-B1596A 50mg Medchemexpress, Ceftizoxime sodium CAS:68401-82-1 Purity:>98%
SDP
Catalog No. 501932975
Encompass_Preferred
Medchemexpress, HY-B1596A 50mg Ceftizoxime sodium CAS:68401-82-1 Ceftizoxime sodium (SKF-88373) is third generation cephalosporin effective against Gram-negative and Gram-positive bacteria. It binds penicillin-binding proteins (PBPs) and inhibits the bacterial cell wall synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2975 Supplier Medchemexpress LLC Supplier No. HYB1596A50MG
Medchemexpress, HY-B1596A 50mg Ceftizoxime sodium CAS:68401-82-1 Ceftizoxime sodium (SKF-88373) is third generation cephalosporin effective against Gram-negative and Gram-positive bacteria. It binds penicillin-binding proteins (PBPs) and inhibits the bacterial cell wall synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.