Learn More
Cayman Chemical Ceftizoxime (sodium salt), Purity: ≥98%, CAS: 68401-82-1, 50 mg
SDP
Catalog No. 502909168
Encompass_Preferred
A cephalosporin antibiotic; active against clinical isolates of the Gram-positive bacteria S. aureus, S. pyogenes, and S. epidermidis (IC50s = 1.56, ≤0.025, and 0.78 µg/ml, respectively), as well as clinical isolates of the Gram-negative bacteria E. coli, K. pneumoniae, and P. mirabilis (IC50s = 0.1, ≤0.025, and ≤0.025 µg/ml, respectively); protective against S. aureus, E. coli, P. mirabilis, or P. vulgaris infection in mice (ED50s = 4.03, 0.02, 0.255, and 0.038 mg/kg, respectively)
Catalog No. 50-290-9168 Supplier Cayman Chemical Supplier No. 3609350 MG
A cephalosporin antibiotic; active against clinical isolates of the Gram-positive bacteria S. aureus, S. pyogenes, and S. epidermidis (IC50s = 1.56, ≤0.025, and 0.78 µg/ml, respectively), as well as clinical isolates of the Gram-negative bacteria E. coli, K. pneumoniae, and P. mirabilis (IC50s = 0.1, ≤0.025, and ≤0.025 µg/ml, respectively); protective against S. aureus, E. coli, P. mirabilis, or P. vulgaris infection in mice (ED50s = 4.03, 0.02, 0.255, and 0.038 mg/kg, respectively)