Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ceftobiprole medocaril sodium | 252188-71-9 | 95.7% | 712.64 g/mol | C26H25N8NaO11S2 | 10 MG
SDP

Catalog No. 5000240447
Encompass_Preferred

Ceftobiprole medocaril sodium is the parenteral prodrug (sodium salt) of ceftobiprole, a pyrrolidinone cephalosporin antibiotic supplied for research use. It displays broad-spectrum in vitro antibacterial activity and is provided as an off-white to light yellow solid for biochemical and antimicrobial studies.

  • Parenteral prodrug sodium salt suitable for in vitro research.
  • Broad-spectrum activity including MRSA and penicillin-resistant streptococci.
  • Purity approximately 95.7% with solid appearance.
  • Molecular weight 712.64 g/mol; formula C26H25N8NaO11S2.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-40447 Supplier Medchemexpress LLC Supplier No. HY106574A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ceftobiprole medocaril sodium is the parenteral prodrug (sodium salt) of ceftobiprole, a pyrrolidinone cephalosporin antibiotic supplied for research use. It displays broad-spectrum in vitro antibacterial activity and is provided as an off-white to light yellow solid for biochemical and antimicrobial studies.

  • Parenteral prodrug sodium salt suitable for in vitro research.
  • Broad-spectrum activity including MRSA and penicillin-resistant streptococci.
  • Purity approximately 95.7% with solid appearance.
  • Molecular weight 712.64 g/mol; formula C26H25N8NaO11S2.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.