Learn More
Medchemexpress LLC Ceftolozane sulfate | 936111-69-2 | 99.7% | 764.77 | 1 MG
SDP
Catalog No. 5000401674
Encompass_Preferred
Ceftolozane sulfate is an antipseudomonal cephalosporin that binds to *P. aeruginosa* essential PBPs (1b, 1c, 2 and 3) with high affinity, inhibiting cell wall synthesis. It is effective against *P. aeruginosa* and Enterobacteriaceae.
  • Inhibits *P. aeruginosa* PAO1 with an MIC of 0.5 μg/mL
  • Effective against AmpC-derepressed *P. aeruginosa*
  • Exhibits antipseudomonal activity against carbapenem-resistant and multi-drug resistant isolates
  • Reduces ESBL-producing Enterobacteriaceae in vivo
  • Reduces bacterial load and improves pulmonary inflammatory response in *P. aeruginosa* pneumonia models
Catalog No. 50-004-01674 Supplier Medchemexpress LLC Supplier No. HY19806A1MG
Ceftolozane sulfate is an antipseudomonal cephalosporin that binds to *P. aeruginosa* essential PBPs (1b, 1c, 2 and 3) with high affinity, inhibiting cell wall synthesis. It is effective against *P. aeruginosa* and Enterobacteriaceae.
  • Inhibits *P. aeruginosa* PAO1 with an MIC of 0.5 μg/mL
  • Effective against AmpC-derepressed *P. aeruginosa*
  • Exhibits antipseudomonal activity against carbapenem-resistant and multi-drug resistant isolates
  • Reduces ESBL-producing Enterobacteriaceae in vivo
  • Reduces bacterial load and improves pulmonary inflammatory response in *P. aeruginosa* pneumonia models