Learn More
Medchemexpress LLC Ceftolozane TFA | 1628046-32-1 | 98.2% | 780.71 | C25H31F3N12O10S2 | 5 MG
SDP
Catalog No. 5000243746
Encompass_Preferred
Ceftolozane TFA is the trifluoroacetic acid (TFA) salt of a cephalosporin antibiotic supplied as a solid for research use. It is used to study and inhibit Gram-negative bacterial infections and as a synthetic building block for development of new antibiotic derivatives.
  • Cephalosporin-class antibiotic active against Gram-negative bacteria.
  • Useful as a synthetic intermediate for antibiotic development.
  • High purity (98.2%) suitable for analytical and synthetic work.
  • White to off-white solid form for easy handling and storage.
  • Available in milligram-scale quantities for research applications.
  • Store at -20°C sealed; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-43746 Supplier Medchemexpress LLC Supplier No. HY106257A5MG
Ceftolozane TFA is the trifluoroacetic acid (TFA) salt of a cephalosporin antibiotic supplied as a solid for research use. It is used to study and inhibit Gram-negative bacterial infections and as a synthetic building block for development of new antibiotic derivatives.
  • Cephalosporin-class antibiotic active against Gram-negative bacteria.
  • Useful as a synthetic intermediate for antibiotic development.
  • High purity (98.2%) suitable for analytical and synthetic work.
  • White to off-white solid form for easy handling and storage.
  • Available in milligram-scale quantities for research applications.
  • Store at -20°C sealed; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • For research use only; not for human or veterinary use.