Learn More
Medchemexpress LLC Ceftolozane TFA | 1628046-32-1 | 98.2% | 780.71 g/mol | C25H31F3N12O10S2 | 1 ML
SDP
Catalog No. 5000244401
Encompass_Preferred
Ceftolozane TFA is a cephalosporin-class antibiotic supplied as a 10 mM solution in DMSO (1 mL). It is intended for use in antibacterial research, analytical method development, and as a building block for synthesis of novel antibiotics. Trifluoroacetate salt; molecular formula C25H31F3N12O10S2; molecular weight 780.71 g/mol.
  • Provided as a ready-to-use 10 mM solution in DMSO.
  • Suitable for antibacterial activity assays and analytical method development.
  • Useful as a synthetic intermediate and reference standard for research.
  • High purity suitable for analytical and preparative applications.
  • Supplied in a 1 mL vial for small-scale experiments.
Catalog No. 50-002-44401 Supplier Medchemexpress LLC Supplier No. HY106257A10MM 1ML
Ceftolozane TFA is a cephalosporin-class antibiotic supplied as a 10 mM solution in DMSO (1 mL). It is intended for use in antibacterial research, analytical method development, and as a building block for synthesis of novel antibiotics. Trifluoroacetate salt; molecular formula C25H31F3N12O10S2; molecular weight 780.71 g/mol.
  • Provided as a ready-to-use 10 mM solution in DMSO.
  • Suitable for antibacterial activity assays and analytical method development.
  • Useful as a synthetic intermediate and reference standard for research.
  • High purity suitable for analytical and preparative applications.
  • Supplied in a 1 mL vial for small-scale experiments.