Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftriaxone Free Acid, G-Biosciences
SCI_ED

Catalog No. S666973
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S666973 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S666973 Supplier G-Biosciences Supplier No. RC955
Add to Cart
Add to Cart

Ceftriaxone free acid is a Beta-lactam cephalosporin compound known for its potent antibiotic activity against gram negative and gram positive bacteria.

It also has been observed to cross the blood brain barrier and has been used for both antibiotic and investigative purposes for the central nervous system (CNS).

Chemical Identifiers

CAS 73384-59-5
Molecular Weight (g/mol) 554.58 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity 98%
Grade Laboratory
Chemical Name or Material Ceftriaxone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.