Learn More
Apexbio Technology LLC Ceftriaxone Sodium Trihydrate 104376-79-6 50mg
SDP
Catalog No. 502240594
Encompass_Preferred
Ceftriaxone Sodium Trihydrate (104376-79-6) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit PBPs thereby disrupting peptidoglycan cross-link formation and compromising bacterial cell wall biosynthesis Ceftriaxone Sodium Trihydrate exerts its biological activity primarily through inhibition of bacterial cell wall synthesis In in vitro studies Ceftriaxone Sodium Trihydrate demonstrates broad-spectrum antimicrobial inhibitory activity with reported IC50 values typically ranging from approximately 0 1 to 4 g/mL depending on the bacterial strain and experimental conditions Based on these pharmacological properties Ceftriaxone Sodium Trihydrate holds research potential in microbiological assays including susceptibility testing drug-resistance research and mechanistic studies involving bacterial PBPs
This item is not returnable. View return policy
Catalog No. 50-224-0594 Supplier Apexbio Technology LLC Supplier No. B169150
Ceftriaxone Sodium Trihydrate (104376-79-6) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit PBPs thereby disrupting peptidoglycan cross-link formation and compromising bacterial cell wall biosynthesis Ceftriaxone Sodium Trihydrate exerts its biological activity primarily through inhibition of bacterial cell wall synthesis In in vitro studies Ceftriaxone Sodium Trihydrate demonstrates broad-spectrum antimicrobial inhibitory activity with reported IC50 values typically ranging from approximately 0 1 to 4 g/mL depending on the bacterial strain and experimental conditions Based on these pharmacological properties Ceftriaxone Sodium Trihydrate holds research potential in microbiological assays including susceptibility testing drug-resistance research and mechanistic studies involving bacterial PBPs