Learn More
Medchemexpress LLC Ceftriaxone sodium salt | 74578-69-1 | 99.9% | 500 MG
SDP
Catalog No. 5000405326
Encompass_Preferred
Ceftriaxone sodium salt (Ro 13-9904) is a broad-spectrum β-lactam third-generation cephalosporin antibiotic effective against various gram-negative and positive bacteria. It acts as a covalent inhibitor of GSK3β with an IC50 of 0.78 μM and also inhibits Aurora B. This compound demonstrates anti-inflammatory, antitumor, and antioxidant activities, making it suitable for bacterial infection and meningitis research.
  • Broad-spectrum β-lactam third-generation cephalosporin antibiotic
  • Effective against various gram-negative and positive bacteria
  • Covalent inhibitor of GSK3β (IC50 value of 0.78 μM)
  • Inhibits Aurora B
  • Exhibits anti-inflammatory, antitumor, and antioxidant activities
  • Suitable for bacterial infection and meningitis research
Catalog No. 50-004-05326 Supplier Medchemexpress LLC Supplier No. HYB0712B500MG
Ceftriaxone sodium salt (Ro 13-9904) is a broad-spectrum β-lactam third-generation cephalosporin antibiotic effective against various gram-negative and positive bacteria. It acts as a covalent inhibitor of GSK3β with an IC50 of 0.78 μM and also inhibits Aurora B. This compound demonstrates anti-inflammatory, antitumor, and antioxidant activities, making it suitable for bacterial infection and meningitis research.
  • Broad-spectrum β-lactam third-generation cephalosporin antibiotic
  • Effective against various gram-negative and positive bacteria
  • Covalent inhibitor of GSK3β (IC50 value of 0.78 μM)
  • Inhibits Aurora B
  • Exhibits anti-inflammatory, antitumor, and antioxidant activities
  • Suitable for bacterial infection and meningitis research