Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ceftriaxone sodium salt hemiheptahydrate

Catalog No. AC455420010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455420010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455420010 Supplier Thermo Scientific Chemicals Supplier No. 455420010
  • $58.90
    $50.65 / Each of 1

    Save $8.25

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ceftriaxone, CAS # 104376-79-6, sodium salt hemiheptahydrate is the sodium salt of ceftriaxone, a third-generation beta-lactam antibiotic of the cephalosporin class.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Ceftriaxone is the sodium salt of ceftriaxone, a beta-lactam antibiotic of the cephalosporin class that works by inactivating penicillin-binding proteins

Applications

• Penicillin-binding protein inactivation inhibits peptidoglycan cross-linkage and leads to destruction of the cell wall and subsequent lysis

• It exhibits antibiotic activity against gram-negative bacteria

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104376-79-6
Molecular Formula C36H46N16Na4O21S6
Molecular Weight (g/mol) 1323.17
MDL Number MFCD01750405,MFCD01750405,MFCD09749665
InChI Key RGDBKADCOSIOEV-MAODNAKNSA-J
PubChem CID 131843450
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Ceftriaxone Sodium Salt Hemiheptahydrate
Melting Point 236.0°C
Infrared Spectrum Authentic
Water 10% max.
Quantity 1 g
Solubility Information Solubility in water: soluble.
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(2-methyl-5,6-dioxo-1H-1,2,4-triazin-3-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium;heptahydrate
Formula Weight 661.6
Percent Purity ≥94% (HPLC)
Chemical Name or Material Ceftriaxone sodium salt hemiheptahydrate
Hazard Category Danger
Hazard Statement GHS H Statement Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.