Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftriaxone Sodium, USP Grade, G-Biosciences
SCI_ED

Catalog No. S675101
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S675101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S675101 Supplier G-Biosciences Supplier No. RC956
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ceftriaxone is a third generation cephalosporin antibiotic, which targets gram-negative and gram-positive bacteria.

It has been used in proteomics research and to study prophylactic strategies to treat hemolytic anemia caused by prescription medications. In addition, ceftriaxone has been used to study the penicillin binding proteins, especially PBP1.

Chemical Identifiers

CAS 104376-79-6
Molecular Weight (g/mol) 661.60 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, GC-MS, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, ASTM 665
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity >95%
Grade USP
Chemical Name or Material Ceftriaxone Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.