Learn More
Selleck Chemical LLC Cefuroxime E4838-5MG
SDP
Catalog No. 5000041942
Encompass_Preferred
Cefuroxime (Kefurox) is an orally active second-generation broad-spectrum cephalosporin antibiotic with increased stability to -lactamase Cefuroxime exhibits broad-spectrum activity against Gram-positive bacteria including penicillinase-producing staphylococci and non- -lactamase-producing gram-negative bacteria
Catalog No. 50-000-41942 Supplier Selleck Chemical LLC Supplier No. E48385MG
Cefuroxime (Kefurox) is an orally active second-generation broad-spectrum cephalosporin antibiotic with increased stability to -lactamase Cefuroxime exhibits broad-spectrum activity against Gram-positive bacteria including penicillinase-producing staphylococci and non- -lactamase-producing gram-negative bacteria