Learn More
Medchemexpress LLC HY-B1325 25mg Medchemexpress, Cefuroxime axetil CAS:64544-07-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB132525MG
Encompass_Preferred
Medchemexpress, HY-B1325 25mg Cefuroxime axetil CAS:64544-07-6 Cefuroxime Axetil, a prodrug of the cephalosporin cefuroxime and an oarl broad spectrum antibiotic, inhibits several gram-positive and gram-negative organisms, including those most frequently associated with various common community-acquired infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1181