Learn More
MOLECULAR DEPOT LLC Cefuroxime BSA - 1 mg
Catalog No. 5000013460
Encompass_Preferred
Cefuroxime BSA conjugate is formed by conjugating the antibiotic cefuroxim to bovine serum albumin This conjugate enhances the immunogenicity of cefuroxim This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13460 Supplier MOLECULAR DEPOT LLC Supplier No. B2021214
Cefuroxime BSA conjugate is formed by conjugating the antibiotic cefuroxim to bovine serum albumin This conjugate enhances the immunogenicity of cefuroxim This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only