Learn More
Cayman Chemical CefuroxIme-d3 1mg
SDP
Catalog No. 502989452
Encompass_Preferred
An internal standard for the quantification of cefuroxime by GC- or LC-MS
Catalog No. 50-298-9452 Supplier Cayman Chemical Supplier No. 311631 MG
An internal standard for the quantification of cefuroxime by GC- or LC-MS