Learn More
Selleck Chemical LLC Cefuroxime sodium S4620-100mg
SDP
Catalog No. 503110266
Encompass_Preferred
Cefuroxime Sodium is a cephalosporin effective against gram-positive and gram-negative bacteria with resistance to -lactamase
Catalog No. 50-311-0266 Supplier Selleck Chemical LLC Supplier No. S4620100MG
Add to Cart
Add to Cart
Cefuroxime Sodium is a cephalosporin effective against gram-positive and gram-negative bacteria with resistance to -lactamase