Learn More
Medchemexpress LLC Cefuroxime sodium | 56238-63-2 | 99.75% | 5 G
SDP
Catalog No. 5000413117
Encompass_Preferred
Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic. It exhibits broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Orally active second-generation cephalosporin antibiotic.
  • Increased stability to β-lactamase.
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria.
Catalog No. 50-004-13117 Supplier Medchemexpress LLC Supplier No. HYB12565G
Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic. It exhibits broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Orally active second-generation cephalosporin antibiotic.
  • Increased stability to β-lactamase.
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria.