Learn More
Medchemexpress LLC Cefuroxime sodium | 56238-63-2 | 99.8% | 1 ML
SDP
Catalog No. 5000413249
Encompass_Preferred
Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic with increased stability to β-lactamase. It exhibits broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Orally active second-generation cephalosporin antibiotic
  • Increased stability to β-lactamase
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria
  • For research use only
Catalog No. 50-004-13249 Supplier Medchemexpress LLC Supplier No. HYB125610MM/1ML
Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic with increased stability to β-lactamase. It exhibits broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Orally active second-generation cephalosporin antibiotic
  • Increased stability to β-lactamase
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria
  • For research use only