Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefuroxime Sodium, G-Biosciences
SCI_ED

Catalog No. S000029
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000029 5 g
S000028 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000029 Supplier G-Biosciences Supplier No. RC958
Add to Cart
Add to Cart

Cefuroxime is a second generation cephalosporin antibiotic targeting gram-positive and gram-negative bacteria. It exhibits some resistance to Beta-lactamase activity.

Cefuroxime binds to specific proteins within the cell wall called penicillin-binding proteins (PBPs), ultimately leading to cell lysis and bacterial death.

Chemical Identifiers

Molecular Weight (g/mol) 446.37 g/mol

Specifications

Quantity 5 g
Percent Purity 97%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.