Learn More
Research Products International Corp RPI Cefuroxime, Sodium Salt, 5 Grams
SDP
Catalog No. 501977238
Encompass_Preferred
Cefuroxime Sodium Salt is a second-generation cephalosporin antibiotic. Cefuroxime inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This interference disrupts the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cefuroxime is effective against a wide spectrum of bacteria, both Gram-positive and Gram-negative. In microbiology and research settings, it is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.  Additionally, Its inclusion allows for the specific growth of bacteria lacking resistance to cefuroxime.
Catalog No. 50-197-7238 Supplier Research Products International Corp Supplier No. C563005.0
Cefuroxime Sodium Salt is a second-generation cephalosporin antibiotic. Cefuroxime inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This interference disrupts the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cefuroxime is effective against a wide spectrum of bacteria, both Gram-positive and Gram-negative. In microbiology and research settings, it is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.  Additionally, Its inclusion allows for the specific growth of bacteria lacking resistance to cefuroxime.