Learn More
Alkali Scientific Cefuroxime, Sodium Salt, 20 G
SDP
Supplier:  Alkali Scientific FUR001CP
Encompass_Preferred
Cefuroxime Sodium Salt, a broad-spectrum cephalosporin antibiotic, is used for the treatment of various bacterial infections. This 20g powder formulation is intended for antibacterial use, particularly in clinical settings for treating infections caused by susceptible organisms. Cefuroxime works by inhibiting bacterial cell wall synthesis, thereby preventing bacterial growth and replication. It is effective against both Gram-positive and Gram-negative bacteria and is commonly used for respiratory, urinary tract, and skin infections. This product is ideal for laboratory use and for preparing solutions for clinical treatments. It is available in a convenient 20g quantity, ensuring sufficient supply for medical and research applications.
Catalog No. 50-006-60500