Learn More
Specifications
Specifications
| CAS | 56238-63-2 |
| Linear Formula | C16H15N4 NaO8 S |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 446.37 |
| Formula Weight | 446.37 |
| Grade | Not Available |
| Physical Form | Neat |
| Chemical Name or Material | Cefuroxime Sodium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.