Learn More
MSC TPI Ceiling Mount Kit, Steel, For ACH and IHP Series, 1 ea
Catalog No. 5000507403
Encompass
This gray ceiling mount kit is designed for use with ACH and IHP series air circulators. Constructed from durable steel, this mount provides a secure attachment solution for overhead fan installations in industrial environments.
  • Compatible with ACH and IHP series circulators
  • Durable steel construction
  • UL listed
  • Requires 1-1/2 in. or longer pipe nipple for assembly (not included)
  • Not compatible with fans featuring a yellow powder coat finish
Catalog No. 50-005-07403 Supplier MSC Supplier No. 77990034
This gray ceiling mount kit is designed for use with ACH and IHP series air circulators. Constructed from durable steel, this mount provides a secure attachment solution for overhead fan installations in industrial environments.
  • Compatible with ACH and IHP series circulators
  • Durable steel construction
  • UL listed
  • Requires 1-1/2 in. or longer pipe nipple for assembly (not included)
  • Not compatible with fans featuring a yellow powder coat finish