Learn More
MSC Vikan Condensation Squeegee, 400 mm, Green
Catalog No. 502839492
Encompass
This condensation squeegee is designed to effectively remove condensation from ceilings and pipes during production. It features drain holes that allow condensation to be collected safely in a bottle or drained away through an attached hose, helping to prevent bacteria growth.
  • Effectively removes condensation from ceilings and pipes.
  • Designed to drain rapidly to help prevent bacteria growth.
  • Pairs with any compatible handle and either a hose or a bottle for condensation removal.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance.
  • Color-coded for hygienic zoning plans and HACCP compliance.
  • Dishwasher safe.
  • Autoclave safe.
Catalog No. 50-283-9492 Supplier MSC Supplier No. 47167176
This condensation squeegee is designed to effectively remove condensation from ceilings and pipes during production. It features drain holes that allow condensation to be collected safely in a bottle or drained away through an attached hose, helping to prevent bacteria growth.
  • Effectively removes condensation from ceilings and pipes.
  • Designed to drain rapidly to help prevent bacteria growth.
  • Pairs with any compatible handle and either a hose or a bottle for condensation removal.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance.
  • Color-coded for hygienic zoning plans and HACCP compliance.
  • Dishwasher safe.
  • Autoclave safe.