Learn More
Grainger Ceiling Tile 48 in L 24 in W PK6
Catalog No. 502753496
Encompass_Preferred
The Armstrong Ceiling Tile 2195 is a 2' x 4' mineral fiber ceiling tile that provides sound absorption and noise reduction. It features a fine texture and white color. The tile has a CAC rating of 35 and NRC rating of 0.55. It meets ASTM E1264 classification for Type III, Form 2, Pattern C. This tile is designed to install in standard 15/16" grid systems for drop ceilings. It provides durability and can be cleaned with mild soap and water. The tile weighs approximately 1 lb/sq ft.
Catalog No. 50-275-3496 Supplier Grainger Supplier No. 13Z998
The Armstrong Ceiling Tile 2195 is a 2' x 4' mineral fiber ceiling tile that provides sound absorption and noise reduction. It features a fine texture and white color. The tile has a CAC rating of 35 and NRC rating of 0.55. It meets ASTM E1264 classification for Type III, Form 2, Pattern C. This tile is designed to install in standard 15/16" grid systems for drop ceilings. It provides durability and can be cleaned with mild soap and water. The tile weighs approximately 1 lb/sq ft.