Learn More
Grainger Ceiling Tile 48 in L 24 in W PK8
Catalog No. 502749885
Encompass_Preferred
The Armstrong Ceiling Tile 13Z993 is a 2' x 2' lay-in acoustical ceiling tile made from mineral fiber. It has a fine texture and 0.55 NRC rating for sound absorption. The tile is designed for standard 15/16" grid systems and has square edges. It is available in white color. The tile meets ASTM E1264 classification for acoustical performance and has a Class A fire rating per ASTM E84. It provides durability and easy maintenance for drop ceilings.
Catalog No. 50-274-9885 Supplier Grainger Supplier No. 13Z993
The Armstrong Ceiling Tile 13Z993 is a 2' x 2' lay-in acoustical ceiling tile made from mineral fiber. It has a fine texture and 0.55 NRC rating for sound absorption. The tile is designed for standard 15/16" grid systems and has square edges. It is available in white color. The tile meets ASTM E1264 classification for acoustical performance and has a Class A fire rating per ASTM E84. It provides durability and easy maintenance for drop ceilings.