Learn More
Hach Company CEL Advanced DrinkingWater Kit
Supplier:  Hach Company 2690100
Encompass
Apparatus set for the CEL Advanced Drinking Water Laboratories Part Nos 2688100 and 251235 Includes 125 mL Erlenmeyer flask 250 mL Erlenmeyer flask 50 mL beaker 100 mL graduated cylinder 25 mL graduated mixing cylinder 50 mL graduated mixing cylinder Digital Titrator with delivery tubes and manual
Catalog No. 50-002-57161