Learn More
Cayman Chemical Cel-Based asy ApIgenIn 100uL
SDP
Catalog No. 502896585
Encompass_Preferred
Source Recombinant MERS-CoV C-Terminal His-tagged nucleocapsid protein expressed in insect cells Amino acids 1-413 (full length) MW 46.51 kDa
Catalog No. 50-289-6585 Supplier Cayman Chemical Supplier No. 600472100 UL
Source Recombinant MERS-CoV C-Terminal His-tagged nucleocapsid protein expressed in insect cells Amino acids 1-413 (full length) MW 46.51 kDa