Learn More
Cayman Chemical Cel-Based asy Buffer B10 25mL
SDP
Catalog No. 502896860
Encompass_Preferred
Cell-Based Assay Buffer B (10X) has been tested and formulated to work exclusively with SARS-CoV-2 Spike S1 RBD-ACE2 Binding Cellular Imaging Assay Kit
Catalog No. 50-289-6860 Supplier Cayman Chemical Supplier No. 70197225 ML
Cell-Based Assay Buffer B (10X) has been tested and formulated to work exclusively with SARS-CoV-2 Spike S1 RBD-ACE2 Binding Cellular Imaging Assay Kit