Learn More
Cayman Chemical Cel-Based asy DAz-2 30uL
SDP
Catalog No. 502896455
Encompass_Preferred
An ionizable cationic lipid (pKa 6.75) has been used in the formation of LNPs for the delivery of mRNA and polymer-lipid hybrid nanoparticles for the delivery of plasmid DNA
Catalog No. 50-289-6455 Supplier Cayman Chemical Supplier No. 60032130 UL
An ionizable cationic lipid (pKa 6.75) has been used in the formation of LNPs for the delivery of mRNA and polymer-lipid hybrid nanoparticles for the delivery of plasmid DNA