Learn More
Cayman Chemical Cel-Based asy GlutathIon 3mg
SDP
Catalog No. 502894968
Encompass_Preferred
An IRE1a inhibitor (IC50s 0.2 and 0.02 UM for RNase and kinase activity respectively) selective for IRE1a over Ron and FGFR1 (IC50s 4.4 and 17 UM) as well as a panel of 282 kinases at 10 UM inhibits ER stress-induced increases in the splicing of mRNA encoding XBP1 in eight multiple myeloma cell lines at 8 UM reduces viral replication in tick-borne encephalitis virus-infected SH-SY5Y and SNB-19 cells at 0.5 UM and tick-borne encephalitis virus-infected Caco-2 cells at 1 UM
Catalog No. 50-289-4968 Supplier Cayman Chemical Supplier No. 6003623 MG
An IRE1a inhibitor (IC50s 0.2 and 0.02 UM for RNase and kinase activity respectively) selective for IRE1a over Ron and FGFR1 (IC50s 4.4 and 17 UM) as well as a panel of 282 kinases at 10 UM inhibits ER stress-induced increases in the splicing of mRNA encoding XBP1 in eight multiple myeloma cell lines at 8 UM reduces viral replication in tick-borne encephalitis virus-infected SH-SY5Y and SNB-19 cells at 0.5 UM and tick-borne encephalitis virus-infected Caco-2 cells at 1 UM