Learn More
Cayman Chemical Cel-Based asy Hoechst Dye 50uL
SDP
Catalog No. 502895873
Encompass_Preferred
A dual peptide GCGR and GLP-1R agonist induces cAMP production in BHK-21 cells expressing human GCGR or GLP-1R (EC50s 1.83 and 0.92 pM respectively) reduces body weight and cumulative food intake in a mouse model of diet-induced obesity from 3-5 nmol/kg reduces body weight hepatic fat accumulation and fibrosis and plasma ALT levels in a mouse model of NASH induced by a high-fat high-fructose and high-cholesterol diet
Catalog No. 50-289-5873 Supplier Cayman Chemical Supplier No. 60033250 UL
A dual peptide GCGR and GLP-1R agonist induces cAMP production in BHK-21 cells expressing human GCGR or GLP-1R (EC50s 1.83 and 0.92 pM respectively) reduces body weight and cumulative food intake in a mouse model of diet-induced obesity from 3-5 nmol/kg reduces body weight hepatic fat accumulation and fibrosis and plasma ALT levels in a mouse model of NASH induced by a high-fat high-fructose and high-cholesterol diet