Learn More
Cayman Chemical Cel-Based asy NIle Red Sol 1ea
SDP
Catalog No. 502896101
Encompass_Preferred
An OGA inhibitor (IC50 46 nM) selective for OGA overBhexosaminidase (IC50 >10 UM) and a panel of 277 kinases (IC50s >1 UM for all) increases the levels of O-GlcNAcylated proteins in SH-SY5Y cells (EC50 450 nM)
Catalog No. 50-289-6101 Supplier Cayman Chemical Supplier No. 6000551 EA
An OGA inhibitor (IC50 46 nM) selective for OGA overBhexosaminidase (IC50 >10 UM) and a panel of 277 kinases (IC50s >1 UM for all) increases the levels of O-GlcNAcylated proteins in SH-SY5Y cells (EC50 450 nM)