Learn More
Cayman Chemical Cel-Based asy VerapamIl 50uL
SDP
Catalog No. 502895195
Encompass_Preferred
A polyamine bile amidate has been found in lion but not mouse fecal samples has been used as a standard in the detection of chenodeoxycholyl-putrescine in mammalian intestinal and fecal samples
Catalog No. 50-289-5195 Supplier Cayman Chemical Supplier No. 60037250 UL
A polyamine bile amidate has been found in lion but not mouse fecal samples has been used as a standard in the detection of chenodeoxycholyl-putrescine in mammalian intestinal and fecal samples