Learn More
Cayman Chemical Cell-free Protein Synthesis (E. coli) Kit, 1 ea
SDP
Catalog No. 502945586
Encompass_Preferred
Cell-free protein synthesis (CFPS) is an easy-to-use, rapid, and efficient system that is devoid of membrane-bound barriers but contains the native cellular transcriptional and translational machinery and exogenous resources (e.g., amino acids, nucleotides, and a secondary energy substrate) needed for synthesis of the desired proteins.{71173,71174,71775,71176,71177} The CFPS (E. coli) Kit can produce high levels and different molecular weights of recombinant proteins within several hours and can be used in high-throughput functional genomics and proteomics. In addition, it can provide proteins to drive biochemical and structural biology research. The synthesized protein can be detected by SDS-PAGE, Western blot, and fluorescence-detection size exclusion chromatography (FSEC) and can be isolated from the reaction mixture by affinity purification techniques for further structural and/or functional characterization.
Catalog No. 50-294-5586 Supplier Cayman Chemical Supplier No. 415341 EA
Cell-free protein synthesis (CFPS) is an easy-to-use, rapid, and efficient system that is devoid of membrane-bound barriers but contains the native cellular transcriptional and translational machinery and exogenous resources (e.g., amino acids, nucleotides, and a secondary energy substrate) needed for synthesis of the desired proteins.{71173,71174,71775,71176,71177} The CFPS (E. coli) Kit can produce high levels and different molecular weights of recombinant proteins within several hours and can be used in high-throughput functional genomics and proteomics. In addition, it can provide proteins to drive biochemical and structural biology research. The synthesized protein can be detected by SDS-PAGE, Western blot, and fluorescence-detection size exclusion chromatography (FSEC) and can be isolated from the reaction mixture by affinity purification techniques for further structural and/or functional characterization.