Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 23436MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: CELA3B/1218. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Panc-28 or PANC1 cells. Pancreas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CELA3B protein fragment (aa82-238). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 17kDa.Chromosome: 1p36.12. Gene Symbol: CELA3B. Gene ID: 23436. Ensemble: ENSG00000219073. Uniprot: NP_031378.
Catalog No. 50-188-4703