Learn More
Cayman Chemical Celastramycin A, Purity: ≥97%, CAS: 491600-94-3, 10 mg
SDP
Catalog No. 502955624
Encompass_Preferred
A bioactive compound that inhibits the growth of bacteria and mycobacteria (minimal inhibitory concentration = 0.05 μg/ml); suppresses TNF-α-stimulated IL-8 production in HUVECs (IC50 = 0.01 μg/ml)
Catalog No. 50-295-5624 Supplier Cayman Chemical Supplier No. 1351910 MG
A bioactive compound that inhibits the growth of bacteria and mycobacteria (minimal inhibitory concentration = 0.05 μg/ml); suppresses TNF-α-stimulated IL-8 production in HUVECs (IC50 = 0.01 μg/ml)