Learn More
Medchemexpress LLC HY-13067 100mg , Celastrol Tripterin CAS:34157-83-0 Purity:98%
SDP
Catalog No. 501493412
Encompass_Preferred
Medchemexpress, HY-13067 100mg Celastrol Tripterin CAS:34157-83-0 Formula:C29H38O4 IC50: 2.5 μM (20S proteasome) Purity:98% Celastrol is a proteasome inhibitor, potently and preferentially inhibits the chymotrypsin-like activity of a purified 20S proteasome ( IC 50 =2.5 μM). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3412 Supplier Medchemexpress LLC Supplier No. HY13067100MG
Medchemexpress, HY-13067 100mg Celastrol Tripterin CAS:34157-83-0 Formula:C29H38O4 IC50: 2.5 μM (20S proteasome) Purity:98% Celastrol is a proteasome inhibitor, potently and preferentially inhibits the chymotrypsin-like activity of a purified 20S proteasome ( IC 50 =2.5 μM). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.