Learn More
Apexbio Technology LLC Celastrol 34157-83-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A26045.1
Encompass_Preferred
Celastrol (CAS 34157-83-0) is a triterpene compound isolated from the Chinese medicinal herb Thunder of God Vine It functions as a proteasome inhibitor targeting the proteolytic activities within the 20S and 26S proteasome complexes leading to intracellular accumulation of ubiquitinated substrates including I B- Bax and p27 thereby inducing apoptosis in cancer cell lines such as human prostate cancer (PC-3 and LNCaP) Celastrol also inhibits NF- B signaling decreasing LPS-induced production of inflammatory cytokines (TNF- and IL-1 ) in human PBMCs and augmenting TNF-mediated apoptosis in KBM-5 cells Its anti-inflammatory and pro-apoptotic properties make celastrol relevant in cancer and inflammation research
Catalog No. 50-000-63322