Learn More
Selleck Chemical LLC Celastrol 50mg 34157-83-0 Tripterine
SDP
Supplier:  Selleck Chemical LLC S129050MG
Encompass_Preferred
Celastrol (Tripterine) is a potent proteasome inhibitor for the chymotrypsin-like activity of a purified 20S proteasome with IC50 of 2.5 ?M. Celastrol induces apoptosis and autophagy via the ROS/JNK signaling pathway. Celastrol inhibits dopaminergic neuronal death of Parkinson's disease through activating mitophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-785-3