Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Celastrol, IL-17A (IL-17A Inhibitor)
SDP

Catalog No. NB002074
Change view
Click to view available options
Quantity:
0.01 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002074 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002074 Supplier Novus Biologicals™ Supplier No. NBP2293620.01G

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Used in assays to inhibit IL-17A induction

CAS Number 34157-83-0

Specifications

Formulation Solubility:DMSO: >10 mg/mL assay:>or equal to 98% (HPLC)
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Buffer Solubility:DMSO: >10 mg/mL assay: ≥ 98% (HPLC)
Quantity 0.01 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.