Learn More
Cayman Chemical CelBased asy NBD CholSte 500uL
SDP
Catalog No. 502894293
Encompass_Preferred
A DYRK1A inhibitor (IC50 0.4 nM) selective for DYRK1A over DYRK2 Clk1 Clk2 Clk3 Cdk2 and GSK3B (IC50s 19 7.1 9.4 54 100 and 94 nM respectively) but does inhibit DYRK1B (IC50 2.7 nM) inhibits DYRK1A-induced increases in tau phosphorylation in a cell-based assay (IC50 590 nM)
Catalog No. 50-289-4293 Supplier Cayman Chemical Supplier No. 600441500 UL
A DYRK1A inhibitor (IC50 0.4 nM) selective for DYRK1A over DYRK2 Clk1 Clk2 Clk3 Cdk2 and GSK3B (IC50s 19 7.1 9.4 54 100 and 94 nM respectively) but does inhibit DYRK1B (IC50 2.7 nM) inhibits DYRK1A-induced increases in tau phosphorylation in a cell-based assay (IC50 590 nM)