Learn More
Apexbio Technology LLC Celecoxib, 169590-42-5, MFCD00941298, 100 mg
SDP
Supplier:  Apexbio Technology LLC A1664100
Encompass_Preferred
MF: C17H14F3N3O2S, MW: 381.37, Solubility: 19.1mg/mL in DMSO. Celecoxib is a highly selective inhibitor of cyclooxygenase-2 (COX-2) with IC50 value of 40nM.
Catalog No. 50-101-1941