Learn More
Medchemexpress LLC HY-14398 100mg Medchemexpress, Celecoxib CAS:169590-42-5 Purity:>98%
SDP
Catalog No. 501932701
Encompass_Preferred
Medchemexpress, HY-14398 100mg Celecoxib CAS:169590-42-5 Celecoxib is a selective COX-2 inhibitor with an IC 50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2701 Supplier Medchemexpress LLC Supplier No. HY14398100MG
Medchemexpress, HY-14398 100mg Celecoxib CAS:169590-42-5 Celecoxib is a selective COX-2 inhibitor with an IC 50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.