Learn More
Medchemexpress LLC Celecoxib | 169590-42-5 | 99.9% | 10 G
SDP
Catalog No. 5000354291
Encompass_Preferred
Celecoxib is a selective and BBB-permeable non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM.
  • Selective COX-2 inhibitor
  • BBB-permeable
  • Non-steroidal anti-inflammatory drug (NSAID)
  • IC50 of 40 nM for COX-2
Catalog No. 50-003-54291 Supplier Medchemexpress LLC Supplier No. HY1439810G
Celecoxib is a selective and BBB-permeable non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM.
  • Selective COX-2 inhibitor
  • BBB-permeable
  • Non-steroidal anti-inflammatory drug (NSAID)
  • IC50 of 40 nM for COX-2