Learn More
Medchemexpress LLC HY-14398 1g , Celecoxib CAS:169590-42-5 Purity:>98%
SDP
Catalog No. 502259103
Encompass_Preferred
HY-14398 1g Celecoxib CAS: 169590-42-5 Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9103 Supplier Medchemexpress LLC Supplier No. HY143981G
Add to Cart
Add to Cart
HY-14398 1g Celecoxib CAS: 169590-42-5 Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.