Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Celecoxib-d3 | 544686-18-2 | 99.0% | 384.39 | C17H11D3F3N3O2S | 5 MG
SDP

Catalog No. 5000222656
Encompass_Preferred

Celecoxib-d3 is a deuterium-labeled analytical reference of celecoxib supplied for research and analytical applications. It is intended as an internal standard for quantitative assays, including LC-MS/MS, to improve accuracy and traceability in pharmacokinetic and metabolic studies.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • High chemical purity (99.0%) to ensure reliable analytical performance.
  • Molecular weight 384.39 and formula C17H11D3F3N3O2S for exact mass calculations.
  • Available in small milligram quantities suitable for analytical workflows.
  • Provided with supporting documentation such as datasheet, COA, and SDS.

Catalog No. 50-002-22656 Supplier Medchemexpress LLC Supplier No. HY14398S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Celecoxib-d3 is a deuterium-labeled analytical reference of celecoxib supplied for research and analytical applications. It is intended as an internal standard for quantitative assays, including LC-MS/MS, to improve accuracy and traceability in pharmacokinetic and metabolic studies.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • High chemical purity (99.0%) to ensure reliable analytical performance.
  • Molecular weight 384.39 and formula C17H11D3F3N3O2S for exact mass calculations.
  • Available in small milligram quantities suitable for analytical workflows.
  • Provided with supporting documentation such as datasheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.