Learn More
Medchemexpress LLC Celecoxib-d3 | 544686-18-2 | 99.0% | C17H11D3F3N3O2S | 1 MG
SDP
Catalog No. 5000353935
Encompass_Preferred
Celecoxib-d3 is the deuterium labeled Celecoxib, a selective non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium have the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-53935 Supplier Medchemexpress LLC Supplier No. HY14398S11MG
Celecoxib-d3 is the deuterium labeled Celecoxib, a selective non-steroidal anti-inflammatory drug (NSAID) and a selective COX-2 inhibitor with an IC50 of 40 nM. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium have the potential to affect the pharmacokinetic and metabolic profiles of drugs.