Learn More
Medchemexpress LLC Celecoxib-d7 | 544686-21-7 | 99.6% | C17H7D7F3N3O2S | 500 UG
SDP
Catalog No. 5000356161
Encompass_Preferred
Celecoxib-d7 is the deuterium-labeled form of Celecoxib, a selective non-steroidal anti-inflammatory drug (NSAID) and COX-2 inhibitor with an IC50 of 40 nM. This compound can serve as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes into drug molecules, such as deuteration, is used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-56161 Supplier Medchemexpress LLC Supplier No. HY14398S500UG
Celecoxib-d7 is the deuterium-labeled form of Celecoxib, a selective non-steroidal anti-inflammatory drug (NSAID) and COX-2 inhibitor with an IC50 of 40 nM. This compound can serve as a tracer and internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes into drug molecules, such as deuteration, is used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs